2-Pentadecanone ≥98% Kosher

Synonyms: Pentadecan-2-one

W372404-5KG-K W372404-100G-K W372404-1KG-K
220235-109EA 350.77 USD
220235-109 220235-107 220235-108
2-Pentadecanone ≥98% Kosher
2-Pentadecanone
Formula: C₁₅H₃₀O
MW: 226.4 g/mol
Density: 0.82 g/cm³ (39 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053712
CAS Number: 2345-28-0
EINECS: 219-064-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR