2-Oxobutyric acid ≥97%

Danger

Synonyms: 2-Oxobutanoic acid, 2-Ketobutyric acid, α-Ketobutyric acid

W372307-250G-K W372307-100G-K W372307-1KG-K
220235-105EA 1907.69 USD
220235-105 220235-103 220235-104
2-Oxobutyric acid ≥97%
2-Oxobutyric acid
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.2 g/cm³ (17 °C)
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004164
CAS Number: 600-18-0
EINECS: 209-986-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR