4-Methylthiazole Kosher

W371602-1KG-K W371602-5KG-K W371602-100G-K
220235-088EA 1615.38 USD
220235-088 220235-089 220235-087
4-Methylthiazole Kosher
4-Methylthiazole
Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 133 °C (1013 hPa)
Density: 1.1187 g/cm³ (20 °C)
MDL Number: MFCD00005340
CAS Number: 693-95-8
EINECS: 211-764-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR