2,6-Dimethoxy-p-cresol ≥97%

Synonyms: 3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol

W370401-1KG W370401-25G
220235-068EA 4261.54 USD
220235-068 220235-069
2,6-Dimethoxy-p-cresol ≥97%
2,6-Dimethoxy-p-cresol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00017289
CAS Number: 6638-05-7
EINECS: 229-641-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR