Hexyl benzoate ≥98%

Synonyms: Benzoic acid hexyl ester

W369101-5KG-K W369101-1KG-K W369101-10KG-K
220235-045EA 650.77 USD
220235-045 220235-044 220235-043
Hexyl benzoate ≥98%
Hexyl benzoate
Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Boiling Pt: 272 °C (1013 hPa)
Density: 0.9992 g/cm³ (0 °C)
MDL Number: MFCD00051714
CAS Number: 6789-88-4
EINECS: 229-856-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR