4-Allyl-2,6-dimethoxyphenol ≥90%

W365505-100G W365505-1KG W365505-25G
220234-977EA 1769.23 USD
220234-977 220234-978 220234-979
4-Allyl-2,6-dimethoxyphenol ≥90%
4-Allyl-2,6-dimethoxyphenol
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00008655
CAS Number: 6627-88-9
EINECS: 229-600-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR