3-Methyl-2-buten-1-ol ≥98%

Synonyms: 2-Methyl-2-buten-4-ol

W364703-4KG-K W364703-1KG-K
220233-599EA 312.31 USD
220233-599 220233-598
3-Methyl-2-buten-1-ol ≥98%
3-Methyl-2-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.848 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR