Dibenzyl disulfide ≥98% Kosher

Warning

Synonyms: Benzyl disulphide

W361704-1KG-K W361704-5KG-K W361704-10KG-K
220233-051EA 100.92 USD
220233-051 220233-052 220233-050
Dibenzyl disulfide ≥98% Kosher
Dibenzyl disulphide
Formula: C₁₄H₁₄S₂
MW: 246.4 g/mol
Boiling Pt: 142…148 °C (0.05 mmHg)
Melting Pt: 62…67 °C
Density: 1.302 g/cm³ (25 °C)
MDL Number: MFCD00004783
CAS Number: 150-60-7
EINECS: 205-764-0
Merck Index: 13,03037

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR