2-Acetyl-5-methylfuran

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

W360902-1KG-K W360902-25G-K W360902-100G-K
220234-893EA 570.77 USD
220234-893 220234-894 220234-892
2-Acetyl-5-methylfuran
2-Acetyl-5-methylfuran
Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00003243
CAS Number: 1193-79-9
EINECS: 214-779-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR