3,4-Xylenol ≥98%

Danger

Synonyms: 4-Hydroxy-2-xylene, 3,4-Dimethylphenol

W359602-250G W359602-1KG W359602-5KG
220234-884EA 172.31 USD
220234-884 220234-883 220234-885
3,4-Xylenol ≥98%
3,4-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 62…65 °C
Density: 1.023 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002304
CAS Number: 95-65-8
EINECS: 202-439-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR