(±)-3-Octanol ≥97% FCC

Warning

Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

W358118-100G-K W358118-1KG-K W358118-8KG-K
220234-840EA 142.92 USD
220234-840 220234-841 220234-842
(±)-3-Octanol ≥97% FCC
(±)-3-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: -45 °C
Density: 0.819 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR