α-Terpinene, FG

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

W355801-8KG-K W355801-4KG-K W355801-1KG-K
220234-798EA 680 USD
220234-798 220234-797 220234-796
α-Terpinene, FG
α-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.834 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00001534
CAS Number: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR