(±)-3-Heptanol ≥98%

Synonyms: 3-Heptyl alcohol, 3-Heptanol, Heptan-3-ol, Butylethylcarbinol, (±)-Ethyl butyl carbinol, (±)-Butyl ethyl carbinol, (±)-3-Hydroxy heptane

W354708-1KG W354708-4KG W354708-100G
220234-772EA 80 USD
220234-772 220234-773 220234-771
(±)-3-Heptanol ≥98%
(±)-3-Heptanol
Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.837 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004586
CAS Number: 589-82-2
EINECS: 209-661-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR