2,6-Lutidine, halal

Synonyms: 2,6-Dimethylpyridine

W354007-5KG-K W354007-10KG-K W354007-1KG-K
220234-761EA 1458.46 USD
220234-761 220234-759 220234-760
2,6-Lutidine, halal
2,6-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 143.7 °C (1013 hPa)
Melting Pt: -6 °C
Density: 0.9252 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III
Merck Index: 13,05636

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR