Pyrrolidine

Danger

Synonyms: Tetrahydropyrrole

W352316-8KG W352316-1KG
220234-727EA 709.23 USD
220234-727 220234-726
Pyrrolidine
Pyrrolidine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 87 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3,II
Merck Index: 12,08199

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR