1,8-Octanedithiol ≥97% Kosher

Warning

Synonyms: 1,8-Dimercaptooctane

W351407-25G-K W351407-100G-K
220231-439EA 640 USD
220231-439 220231-437
1,8-Octanedithiol ≥97% Kosher
1,8-Octanedithiol
Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 270 °C (1013 hPa)
Density: 0.9814 g/cm³ (0 °C)
MDL Number: MFCD00003574
CAS Number: 1191-62-4
EINECS: 214-738-8
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR