2-Methyl-4-pentenoic acid ≥98% Kosher

Synonyms: 2-Methylpent-4-enoic acid

W351105-100G-K W351105-5KG-K
220234-703EA 80 USD
220234-703 220234-705
2-Methyl-4-pentenoic acid ≥98% Kosher
2-Methyl-4-pentenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 188 - 189 °C (683 -rr)
Density: 0.95 g/cm³ (20 °C)
MDL Number: MFCD00015570
CAS Number: 1575-74-2
EINECS: 216-404-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR