2-Methylbutyl isovalerate Kosher

Synonyms: Isopropyl acetic acid 2-methylbutyl ester, Isovaleric acid 2-methylbutyl ester, 2-Methylbutyl isopentanoate, 3-Methylbutyric acid 2-methylbutyl ester, 3-Methylbutanoic acid 2-methylbutyl ester

W350613-4KG-K W350613-100G-K W350613-1KG-K
220234-689EA 373.85 USD
220234-689 220234-687 220234-688
2-Methylbutyl isovalerate Kosher
2-Methylbutyl isovalerate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
CAS Number: 2445-77-4
EINECS: 219-496-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR