2-Methylbutyl isovalerate ≥98% Kosher

Synonyms: Isopropyl acetic acid 2-methylbutyl ester, Isovaleric acid 2-methylbutyl ester, 2-Methylbutyl isopentanoate, 3-Methylbutyric acid 2-methylbutyl ester, 3-Methylbutanoic acid 2-methylbutyl ester

W350605-1KG-K W350605-4KG-K W350605-9KG-K
220234-683EA 232.31 USD
220234-683 220234-684 220234-685
2-Methylbutyl isovalerate ≥98% Kosher
2-Methylbutyl isovalerate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
CAS Number: 2445-77-4
EINECS: 219-496-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR