Ethyl stearate ≥97%

Synonyms: Stearic acid ethyl ester, Ethyl octadecanoate

W349003-1KG W349003-250G W349003-5KG
220234-658EA 180 USD
220234-658 220234-659 220234-660
Ethyl stearate ≥97%
Ethyl stearate
Formula: C₂₀H₄₀O₂
MW: 312.54 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 31 °C
Density: 0.8973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009006
CAS Number: 111-61-5
EINECS: 203-887-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR