Ethyl crotonate ≥96% Kosher

Danger

Synonyms: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester

W348600-10KG-K W348600-20KG-K W348600-1KG-K
220234-640EA 2292.31 USD
220234-640 220234-642 220234-641
Ethyl crotonate ≥96% Kosher
Ethyl crotonate
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.9208 g/cm³ (20 °C)
Flash Pt: 2 °C
MDL Number: MFCD00009289
CAS Number: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR