4-Methylvaleric acid ≥98%, FG

Danger

Synonyms: Isocaproic acid, 4-Methylpentanoic acid

W346306-1KG-K W346306-250G-K W346306-10KG-K
220234-601EA 704.62 USD
220234-601 220234-602 220234-600
4-Methylvaleric acid ≥98%, FG
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: -35 °C
Density: 0.9192 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR