2-Isopropylphenol ≥98%

Danger

W346101-10KG W346101-25KG W346101-1KG
220234-592EA 2276.92 USD
220234-592 220234-594 220234-593
2-Isopropylphenol ≥98%
2-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 14 - 16 °C
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR