DL-Valine ≥97%

W344400-10KG W344400-1KG
220234-571EA 1769.23 USD
220234-571 220234-572
DL-Valine ≥97%
DL-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR