(±)-3-Methylvaleric acid ≥98%, FG

Danger

Synonyms: 3-Methylvaleric Acid, 3-Methylpentanoic acid, (±)-3-Methylpentanoic acid

W343706-25G-K W343706-1KG-K W343706-100G-K
220234-058EA 190.77 USD
220234-058 220234-057 220234-056
(±)-3-Methylvaleric acid ≥98%, FG
(±)-3-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: -41 °C
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002729
CAS Number: 105-43-1
EINECS: 203-297-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR