2-Isopropyl-5-methylanisole Kosher

Danger

Synonyms: 4-Isopropyl-3-methoxytoluene, 1-Isopropyl-2-methoxy-4-methylbenzene

W343609-1KG-K W343609-5KG-K W343609-100G-K
220234-053EA 4292.31 USD
220234-053 220234-054 220234-052
2-Isopropyl-5-methylanisole Kosher
2-Isopropyl-5-methylanisole
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
MDL Number: MFCD01674973
CAS Number: 1076-56-8
EINECS: 214-063-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR