3-Methyl-2-cyclopenten-1-one

Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone

W343502-100G W343502-25G
220234-501EA 1984.62 USD
220234-501 220234-502
3-Methyl-2-cyclopenten-1-one
3-Methyl-2-cyclopenten-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 2 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001403
CAS Number: 2758-18-1
EINECS: 220-421-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR