2,4-Dimethylbenzaldehyde ≥90%

W342718-1KG W342718-100G
220234-474EA 4292.31 USD
220234-474 220234-473
2,4-Dimethylbenzaldehyde ≥90%
2,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.96 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003340
CAS Number: 15764-16-6
EINECS: 239-856-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR