Dimethyl acetal ≥97%

Danger

Synonyms: Acetaldehyde dimethyl acetal, 1,1-Dimethoxyethane

W342602-1KG W342602-4KG W342602-100G
220234-470EA 696.92 USD
220234-470 220234-471 220234-469
Dimethyl acetal ≥97%
Dimethyl acetal
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –113 °C
Density: 0.8505 g/cm³ (20 °C)
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3,II
Merck Index: 13,03253

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR