3-Acetylpyridine Kosher

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

W342408-1KG-K W342408-5KG-K W342408-100G-K W342408-10KG-K
220234-465EA 726.15 USD
220234-465 220234-467 220234-463 220234-464
3-Acetylpyridine Kosher
3-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220 °C (1013 hPa)
Density: 1.1065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR