trans-2-Undecenal ≥90% Kosher

Synonyms: (E)-2-Undecenal

W342300-1KG-K W342300-100G-K W342300-4KG-K
220234-460EA 1407.69 USD
220234-460 220234-459 220234-461
trans-2-Undecenal ≥90% Kosher
trans-2-Undecenal
Formula: C₁₁H₂₀O
MW: 168.28 g/mol
MDL Number: MFCD00014680
CAS Number: 53448-07-0
EINECS: 258-559-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR