trans-2-Nonen-1-ol ≥96% Kosher

Synonyms: (E)-2-Nonen-1-ol

W337900-1KG-K W337900-25G-K W337900-100G-K
220234-321EA 80 USD
220234-321 220234-322 220234-320
trans-2-Nonen-1-ol ≥96% Kosher
trans-2-Nonen-1-ol
Formula: C₉H₁₈O
MW: 142.24 g/mol
MDL Number: MFCD00014058
CAS Number: 31502-14-4
EINECS: 250-662-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR