Mesityl oxide ≥90%

Danger

Synonyms: 4-Methyl-3-penten-2-one

W336807-9KG W336807-1KG W336807-4KG
220234-293EA 515.38 USD
220234-293 220234-291 220234-292
Mesityl oxide ≥90%
Mesityl oxide
Formula: C₆H₁₀O
MW: 98.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008900
CAS Number: 141-79-7
EINECS: 205-502-5
UN: 1229
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR