3-Methyl-2-cyclohexen-1-one ≥98%

Synonyms: 3-Methyl-2-cyclohexenone, 3-Methylcyclohex-2-enone

W336009-100G-K W336009-1KG-K
220232-921EA 80 USD
220232-921 220232-922
3-Methyl-2-cyclohexen-1-one ≥98%
3-Methyl-2-cyclohexen-1-one
Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 198 - 199 °C
Density: 0.9791 g/cm³ (10 °C)
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR