Furfuryl propionate, halal

Synonyms: Furan-2-ylmethyl Propionate, Propionoic acid Furfuryl ester, Propionoic acid Furan-2-ylmethyl ester

W334618-1KG-K W334618-5KG-K W334618-100G-K
220234-281EA 524.62 USD
220234-281 220233-537 220234-280
Furfuryl propionate, halal
Furfuryl propionate
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD00040262
CAS Number: 623-19-8
EINECS: 210-776-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR