2-Acetylthiazole, halal

Synonyms: 1-(Thiazol-2-yl)-1-ethanone

W332801-1KG-K W332801-5KG-K W332801-100G-K
220234-255EA 1815.38 USD
220234-255 220234-256 220234-254
2-Acetylthiazole, halal
2-Acetylthiazole
Formula: C₅H₅NOS
MW: 127.17 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 65.5 °C
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00005324
CAS Number: 24295-03-2
EINECS: 246-134-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR