L(-)-Proline FCC

Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH

W331902-1KG-K W331902-100G-K W331902-5KG-K
220234-229EA 227.69 USD
220234-229 220234-228 220234-230
L(-)-Proline FCC
L(-)-Proline
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR