(±)-2-Pentanol ≥98% Kosher

Danger

Synonyms: (±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol

W331600-1KG-K W331600-4KG-K
220234-220EA 189.23 USD
220234-220 220234-221
(±)-2-Pentanol ≥98% Kosher
(±)-2-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 12,07258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR