(±)-2-Nonanol ≥97%

Synonyms: (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol

W331503-100G W331503-1KG
220234-216EA 278.46 USD
220234-216 220234-217
(±)-2-Nonanol ≥97%
(±)-2-Nonanol
Formula: C₉H₂₀O
MW: 144.26 g/mol
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.831 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004593
CAS Number: 628-99-9
EINECS: 211-065-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR