3-(Methylthio)propyl isothiocyanate, FG

Synonyms: 3-(Methylthio)propylisothiocyanate

W331201-1KG-K W331201-100G-K W331201-25G-K
220232-557EA 1358.46 USD
220232-557 220232-556 220231-637
3-(Methylthio)propyl isothiocyanate, FG
3-(Methylthio)propyl isothiocyanate
Formula: C₅H₉NS₂
MW: 147.27 g/mol
MDL Number: MFCD00209521
CAS Number: 505-79-3
EINECS: 208-020-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR