L(+)-Leucine ≥98.5% Kosher

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

W329703-1KG-K W329703-5KG-K W329703-100G-K
220232-936EA 215.38 USD
220232-936 220232-937 220232-935
L(+)-Leucine ≥98.5% Kosher
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 300 °C
Density: 1.165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR