DL-Isoleucine ≥98.5% FCC

Synonyms: H-DL-Ile-OH

W329509-25G W329509-100G
220232-904EA 164.62 USD
220232-904 220232-902
DL-Isoleucine ≥98.5% FCC
DL-Isoleucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 275 °C
Density: 1.255 g/cm³ (-123.15 °C)
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8
Merck Index: 13,05198

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR