(±)-δ-Undecalactone Kosher

Synonyms: delta-Undecanolactone, (±)-Tetrahydro-6-hexyl-2H-pyran-2-one, 5-Hydroxyundecanoic acid delta-Lactone, (±)-Undecan-5-olide, Undecanoic δ-lactone, 6-Hexyltetrahydro-2H-pyran-2-one

W329401-1KG-K W329401-5KG-K W329401-100G-K
220232-899EA 544.62 USD
220232-899 220232-900 220232-898
(±)-δ-Undecalactone Kosher
(±)-δ-Undecalactone
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
MDL Number: MFCD00006650
CAS Number: 710-04-3
EINECS: 211-915-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR