α-Angelicalactone, FG

Synonyms: 5-Methylfuran-2(3H)-one

W329304-1KG-K W329304-250G-K W329304-100G-K
220232-895EA 1303.08 USD
220232-895 220232-896 220234-110
α-Angelicalactone, FG
α-Angelicalactone
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.0904 g/cm³ (12 °C)
MDL Number: MFCD00005375
CAS Number: 591-12-8
EINECS: 209-701-8
Merck Index: 12,00686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR