3-Hexanone ≥97%

Danger

W329002-100G W329002-25G W329002-1KG
220234-102EA 187.69 USD
220234-102 220234-104 220234-103
3-Hexanone ≥97%
3-Hexanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: –55.4 °C
Density: 0.8216 g/cm³ (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009402
CAS Number: 589-38-8
EINECS: 209-645-4
UN: 1224
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR