Disodium succinate ≥98%, halal

Synonyms: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic

W327700-5KG-K W327700-1KG-K W327700-10KG-K
220234-017EA 543.08 USD
220234-017 220234-016 220234-015
Disodium succinate ≥98%, halal
Disodium succinate
Formula: C₄H₄Na₂O₄
MW: 162.05 g/mol
Density: 1.88 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002790
CAS Number: 150-90-3
EINECS: 205-778-7
Merck Index: 12,08828

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR