4,5-Dimethylthiazole

W327409-100G W327409-1KG W327409-25G
220234-007EA 446.15 USD
220234-007 220234-008 220234-009
4,5-Dimethylthiazole
4,5-Dimethylthiazole
Formula: C₅H₇NS
MW: 113.18 g/mol
Boiling Pt: 149 - 153 °C (1013 hPa)
Density: 1.0828 g/cm³ (20 °C)
MDL Number: MFCD00005336
CAS Number: 3581-91-7
EINECS: 222-703-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR