2,5-Dimethylpyrazine Kosher

Warning

W327204-100G-K W327204-500G-K W327204-1KG-K
220233-997EA 141.23 USD
220233-997 220233-999 220233-998
2,5-Dimethylpyrazine Kosher
2,5-Dimethylpyrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR