3,5-Dimethyl-1,2-cyclopentanedione ≥97%

Synonyms: 3,5-Dimethylcyclopentane-1,2-dione

W326909-100G-K W326909-25G-K W326909-1KG-K
220233-988EA 863.08 USD
220233-988 220233-990 220233-989
3,5-Dimethyl-1,2-cyclopentanedione ≥97%
3,5-Dimethyl-1,2-cyclopentanedione
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00143076
CAS Number: 13494-07-0
EINECS: 236-811-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR