3,4-Dimethyl-1,2-cyclopentanedione ≥97%, halal

Synonyms: 3,4-Dimethylcyclopentane-1,2-dione, 3,4-Dimethyl-1,2-cyclopentadione

W326801-100G-K W326801-25G-K W326801-1KG-K
220233-982EA 607.69 USD
220233-982 220233-984 220233-983
3,4-Dimethyl-1,2-cyclopentanedione ≥97%, halal
3,4-Dimethyl-1,2-cyclopentanedione
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 71…72 °C
MDL Number: MFCD00143063
CAS Number: 13494-06-9
EINECS: 236-810-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR